Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Galanin Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC5854R100UG
This item is not returnable.
View return policy
Description
Galanin Monoclonal specifically detects Galanin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Galanin | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P22466 | |
GAL | |
E.coli-derived recombinant human Galanin Ala20-Ser123 Accession # P22466 | |
RUO | |
51083 | |
Human | |
IgG2b |
Flow Cytometry | |
581403 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
GAL, GAL1, galanin, galanin prepropeptide, GALN, GALNgalanin-related peptide, GLNNgalanin-message-associated peptide, GMAP, MGC40167 | |
Mouse | |
100 μg | |
Primary | |
Detects human Galanin in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction