Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Galanin R2/GALR2 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6544G100UG
This item is not returnable.
View return policy
Description
Galanin R2/GALR2 Monoclonal specifically detects Galanin R2/GALR2 in Human samples. It is validated for Flow Cytometry.Specifications
Galanin R2/GALR2 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
GAL2R, GAL2-R, galanin receptor 2, galanin receptor type 2, GalaninR2, GALNR2, GALNR2MGC125984, GALR2, GALR-2, MGC125983 | |
NS0 mouse myeloma cell line transfected with human Galanin R2, Accession # O43603 | |
100 μg | |
8811 | |
Human | |
Supernatant |
Flow Cytometry | |
616520 | |
Flow Cytometry 0.25-1 μg/106 cells | |
O43603 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human GAL-R2 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction