Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Galanin R2/GALR2 Alexa Fluor 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6544N10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6544N10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6544N10 Supplier R&D Systems Supplier No. FAB6544N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Galanin R2/GALR2 Monoclonal specifically detects Galanin R2/GALR2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Galanin R2/GALR2
Applications Flow Cytometry
Classification Monoclonal
Clone 616520
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. O43603
Gene Alias GAL2R, GAL2-R, galanin receptor 2, galanin receptor type 2, GalaninR2, GALNR2, GALNR2MGC125984, GALR2, GALR-2, MGC125983
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Galanin R2, Accession # O43603
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 8811
Test Specificity Detects human GAL-R2 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.