Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Galectin-10 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB5447V100UG
This item is not returnable.
View return policy
Description
Galectin-10 Monoclonal specifically detects Galectin-10 in Human samples. It is validated for Flow Cytometry.Specifications
Galectin-10 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q05315 | |
CLC | |
E. coli-derived recombinant human Galectin-10 Met1-Arg142 Accession # Q05315 | |
RUO | |
1178 | |
Human | |
IgG2b |
Flow Cytometry | |
561603 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Charcot-Leyden crystal proteinLGALS10A, CLC, EC 3.1.1.5, eosinophil lysophospholipase, Gal-10, GAL10, galectin 10, galectin-10, Galectin10, LGALS10, LPPL_HUMAN, Lysolecithin acylhydrolase, MGC149659 | |
Mouse | |
100 μg | |
Primary | |
Detects human Galectin-10 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Galectin-1, -2, -3, -4, -7, -8, or -9/Ecalectin is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction