Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Galectin-12 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10157100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10157100 100 μg
MAB10157SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10157100 Supplier R&D Systems Supplier No. MAB10157100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Galectin-12 Monoclonal specifically detects Galectin-12 in Human samples. It is validated for ELISA.

Specifications

Antigen Galectin-12
Applications ELISA
Classification Monoclonal
Clone 984513
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q96DT0
Gene Alias Gal-12, GAL12, galectin 12, Galectin-related inhibitor of proliferation, GRIP1, GRIP1galectin-12, lectin, galactoside-binding, soluble, 12, LGALS12
Host Species Mouse
Immunogen Human Galectin-12 synthetic peptide, Accession # Q96DT0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 85329
Test Specificity Detects human Galectin-12 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.