Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Galectin-2 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC1153G100UG
This item is not returnable.
View return policy
Description
Galectin-2 Monoclonal specifically detects Galectin-2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Galectin-2 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Beta-galactoside-binding lectin L-14-II, Gal-2, GAL2, galectin 2, galectin-2, Galectin2, HL14gal-2, Lactose-binding lectin 2, lectin, galactoside-binding, soluble, 2, LGALS2, MGC75071, S-Lac lectin 2 | |
E. coli-derived human Galectin-2, Met1-Glu132, Accession # P05162 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2509A | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P05162 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
3957 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction