Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Human Galectin-2 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Monoclonal Antibody
Supplier: R&D Systems IC1153N100UG
                        
                            
                        
                        
                        
                            
                        
                        
                        
                        
                            
                                    
                                This item is not returnable.
                                View return policy
                                
                        
                    
                Description
Galectin-2 Monoclonal specifically detects Galectin-2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| Galectin-2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Beta-galactoside-binding lectin L-14-II, Gal-2, GAL2, galectin 2, galectin-2, Galectin2, HL14gal-2, Lactose-binding lectin 2, lectin, galactoside-binding, soluble, 2, LGALS2, MGC75071, S-Lac lectin 2 | |
| E. coli-derived human Galectin-2, Met1-Glu132, Accession # P05162 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified | 
| Flow Cytometry | |
| 2509A | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P05162 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3957 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction