Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Galectin-3 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC11542G100UG
This item is not returnable.
View return policy
Description
Galectin-3 Monoclonal specifically detects Galectin-3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Galectin-3 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P17931 | |
LGALS3 | |
E. coli-derived recombinant human Galectin-3 Ala2-Ile250 Accession # P17931 | |
RUO | |
3958 | |
Human | |
IgG2b |
Flow Cytometry | |
194801 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
AGE-R3, Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, CBP35, Gal-3, GAL3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, Galectin3, GALIG, IgE-binding protein, L29, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, LGALS3, Mac-2, Mac-2 antigen, MAC2GAL3 | |
Mouse | |
100 μg | |
Primary | |
Detects human Galectin-3 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant mouse Galectin-3 is observed and no cross-reactivity with recombinant human (rh) Galectin-8 or rhGalectin-9 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction