Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Galectin-3 APC-conjugated Antibody, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems IC1154A
This item is not returnable.
View return policy
Description
Galectin-3 Polyclonal specifically detects Galectin-3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Galectin-3 | |
Polyclonal | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
AGE-R3, Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, CBP35, Gal-3, GAL3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, Galectin3, GALIG, IgE-binding protein, L29, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, LGALS3, Mac-2, Mac-2 antigen, MAC2GAL3 | |
Goat | |
Affinity Purified | |
RUO | |
3958 | |
Human | |
IgG |
Flow Cytometry | |
APC | |
P17931.5 | |
LGALS3 | |
E. coli-derived recombinant human Galectin-3 Ala2-Ile250 Accession # P17931.5 | |
100 Tests | |
Primary | |
Detects human Galectin-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse Galectin-3 is observed and less than 1% cross-reactivity with recombinant human Galectin-1, -2, -4, -7, and -8 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction