Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Galectin-3 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC11542P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC11542P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC11542P Supplier R&D Systems Supplier No. IC11542P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Galectin-3 Monoclonal specifically detects Galectin-3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Galectin-3
Applications Flow Cytometry
Classification Monoclonal
Clone 194801
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P17931
Gene Alias AGE-R3, Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, CBP35, Gal-3, GAL3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, Galectin3, GALIG, IgE-binding protein, L29, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, LGALS3, Mac-2, Mac-2 antigen, MAC2GAL3
Gene Symbols LGALS3
Host Species Mouse
Immunogen E. coli-derived recombinant human Galectin-3 Ala2-Ile250 Accession # P17931
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3958
Test Specificity Detects human Galectin-3 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant mouse Galectin-3 is observed and no cross-reactivity with recombinant human (rh) Galectin-8 or rhGalectin-9 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.