Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Galectin-3BP/MAC-2BP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2226SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2226SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2226SP Supplier R&D Systems Supplier No. AF2226SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Galectin-3BP/MAC-2BP/LGALS3BP Polyclonal specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.

Specifications

Antigen Galectin-3BP/MAC-2BP/LGALS3BP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 1-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q08380
Gene Alias 90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, Galectin3BP, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, LGALS3BP, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K
Gene Symbols LGALS3BP
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Galectin-3BP/MAC-2BP Val19-Asp585 Accession # Q08380
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3959
Test Specificity Detects human Galectin-3BP/MAC-2BP in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Galectin-1, rhGalectin-2, rhGalectin-7, rhGalectin-8, rhGalectin-9, and recombinant mouse (rm) Galectin-1 is observed. In direct ELISAs, approximately 1% cross-reactivity with rmGalectin-9, approximately 8% cross-reactivity with rmGalectin-3, and approximately 15% cross-reactivity with rmMAC-2BP is observed. In Western blots, approximately 15% cross-reactivity with recombinant mouse MAC-2BP is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.