Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Galectin-3BP/MAC-2BP Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2226
This item is not returnable.
View return policy
Description
Galectin-3BP/MAC-2BP/LGALS3BP Polyclonal specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
Galectin-3BP/MAC-2BP/LGALS3BP | |
Polyclonal | |
Unconjugated | |
Q08380 | |
LGALS3BP | |
Mouse myeloma cell line NS0-derived recombinant human Galectin-3BP/MAC-2BP Val19-Asp585 Accession # Q08380 | |
100 μg | |
Primary | |
Detects human Galectin-3BP/MAC-2BP in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Galectin-1, rhGalectin-2, rhGalectin-7, rhGalectin-8, rhGalectin-9, and recombinant mouse (rm) Galectin-1 is observed. In direct ELISAs, approximately 1% cross-reactivity with rmGalectin-9, approximately 8% cross-reactivity with rmGalectin-3, and approximately 15% cross-reactivity with rmMAC-2BP is observed. In Western blots, approximately 15% cross-reactivity with recombinant mouse MAC-2BP is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.2 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 1-15 ug/mL | |
90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, Galectin3BP, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, LGALS3BP, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K | |
Goat | |
Affinity Purified | |
RUO | |
3959 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction