Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Galectin-3BP/MAC-2BP/LGALS3BP ELISA Kit (Colorimetric)
SDP

Catalog No. NBP191179 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP191179 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP191179 Supplier Novus Biologicals™ Supplier No. NBP191179

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Human Galectin-3BP/MAC-2BP/LGALS3BP ELISA Kit (Colorimetric) measures human Galectin-3BP/MAC-2BP/LGALS3BP in cell culture supernatant, serum, plasma.

Specifications

Assay Range 12.5 - 200ng/mL
Assay Sensitivity 0.92ng/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma, Serum
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3959
Gene Symbol LGALS3BP
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with Monoclonal Antibody (murine) to human 90K/Mac-2 BP, HRP-Conjugate anti-90K/Mac-2 BP monoclonal (murine) antibody, 90K/Mac-2 BP Standard, 200ng/ml; 100ng/ml;50ng/ml;25ng/ml; 12.5ng/ml, Wash Buffer Concentrate 10x (phosphate-buffered saline with Tween 20), Sample Diluent, 10x (buffered protein matrix), Substrate Solution (tetramethyl-benzidine and hydrogen peroxide, stabilized), Stop Solution (0.3M Sulfuric Acid), Adhesive Plate Covers, Reagent Labels
Quantity 96 Tests
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym 90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2-B
Target Galectin-3BP/MAC-2BP/LGALS3BP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.