Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Galectin-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1227
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1227 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1227 Supplier R&D Systems Supplier No. MAB1227
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Galectin-4 Monoclonal specifically detects Galectin-4 in Human samples. It is validated for Western Blot.

Specifications

Antigen Galectin-4
Applications Western Blot
Classification Monoclonal
Clone 198616
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P56470
Gene Alias Antigen NY-CO-27, gal-4, GAL4, galectin 4, galectin-4, Galectin4, L-36 lactose-binding protein, L36LBP, Lactose-binding lectin 4, lectin, galactoside-binding, soluble, 4, LGALS4
Gene Symbols LGALS4
Host Species Mouse
Immunogen E. coli-derived recombinant human Galectin-4 Ala2-Ile323 Accession # P56470
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3960
Test Specificity Detects human Galectin-4 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Galectin-1, -2, -3, -7, -8, recombinant mouse Galectin-1, -3, -4, or -7 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.