Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Galectin-7 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA13392T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA13392T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA13392T100 Supplier R&D Systems Supplier No. FAB13392T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Galectin-7 Monoclonal specifically detects Galectin-7 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Galectin-7
Applications Flow Cytometry
Classification Monoclonal
Clone 950723
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. NP_002298
Gene Alias Gal-7, GAL7, galectin 7, galectin-7, Galectin7, HKL-14, lectin, galactoside-binding, soluble, 7, LGALS7, LGALS7A, p53-induced gene 1 protein, PI7, PIG1, TP53I1
Host Species Mouse
Immunogen E. coli-derived recombinant human Galectin-7 Ser2-Phe136 Accession # NP_002298
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3963
Test Specificity Detects human Galectin-7 in direct ELISAs and Western blots.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.