Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Galectin-8 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 12 publications
Supplier: R&D Systems AF1305
This item is not returnable.
View return policy
Description
Galectin-8 Polyclonal specifically detects Galectin-8 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
Galectin-8 | |
Polyclonal | |
Unconjugated | |
O00214 | |
LGALS8 | |
E. coli-derived recombinant human Galectin-8 Met1-Trp317 Accession # O00214 | |
100 μg | |
Primary | |
Detects human Galectin-8 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) rhGalectin-1 and rhGalectin-3 is observed, and less than 1% cross-reactivity with rhGalectin-2, rhGalectin-4 and rhGalectin-7 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Gal-8, GAL8, galectin 8, galectin-8, Galectin8, galectin-8g, lectin, galactoside-binding, soluble, 8, LGALS8, PCTA1, PCTA-1Po66 carbohydrate-binding protein, Po66 carbohydrate binding protein, Po66-CBP, Prostate carcinoma tumor antigen 1 | |
Goat | |
Affinity Purified | |
RUO | |
3964 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction