Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Galectin-9 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$188.70 - $454.00
Specifications
Antigen | Galectin-9 |
---|---|
Clone | 2315B |
Dilution | 0.25 μg/106 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9064100
![]() |
R&D Systems
MAB9064100 |
100 μg |
Each for $454.00
|
|
|||||
MAB9064SP
![]() |
R&D Systems
MAB9064SP |
25 μg |
Each for $188.70
|
|
|||||
Description
Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Human samples. It is validated for Flow CytometrySpecifications
Galectin-9 | |
0.25 μg/106 cells | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
ecalectin, Gal-9, galectin 9, galectin-9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein | |
Human embryonic kidney cell, HEK293 derived human Galectin-9, Met1-Thr323, Accession # O00182 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2315B | |
Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human | |
O00182 | |
3965 | |
IgG | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title