Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Galectin-9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | Galectin-9 |
---|---|
Clone | 1005451 |
Dilution | Western Blot 2 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB20456SP
![]() |
R&D Systems
MAB20456SP |
25 μg |
Each for $200.50
|
|
|||||
MAB20456100
![]() |
R&D Systems
MAB20456100 |
100 μg |
Each for $432.50
|
|
|||||
Description
Galectin-9 Monoclonal specifically detects Galectin-9 in Human samples. It is validated for Western Blot.Specifications
Galectin-9 | |
Western Blot 2 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
ecalectin, gal-9, GAL9, galectin 9, galectin-9, Galectin9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein | |
E. coli-derived human Galectin-9, Ala2-Thr323, Accession # O00182 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1005451 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O00182 | |
3965 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title