Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Galectin-9 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems BAF2045
This item is not returnable.
View return policy
Description
Galectin-9 Polyclonal specifically detects Galectin-9 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Galectin-9 | |
Polyclonal | |
Biotin | |
ecalectin, gal-9, GAL9, galectin 9, galectin-9, Galectin9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein | |
Goat | |
Affinity Purified | |
RUO | |
3965 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
LGALS9 | |
E. coli-derived recombinant human Galectin-9 | |
50 μg | |
Primary | |
Detects human Galectin-9 in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human (rh) Galectin-3 and rhGalectin-4 is observed and less than 2% cross-reactivity with rhGalectin-1, rhGalectin-2, rhGalectin-7, and rhGalectin-8 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction