Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human gamma-Synuclein Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5745
This item is not returnable.
View return policy
Description
gamma-Synuclein Monoclonal specifically detects gamma-Synuclein in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
gamma-Synuclein | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q6FHG5 | |
SNCG | |
E. coli-derived recombinant human gamma-Synuclein Asn47-Glu120 Accession # Q6FHG5 | |
100 μg | |
Primary | |
Detects human gamma-Synuclein in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
514304 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BCSG1, BCSG1PERSYN, Breast cancer-specific gene 1 protein, gamma-synuclein, persyn, PRSN, SNCG, SRPRSN, synoretin, synuclein, gamma (breast cancer-specific protein 1), Synuclein-gamma | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6623 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction