Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Gas1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2636
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2636 500 μg
MAB2636SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2636 Supplier R&D Systems Supplier No. MAB2636
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Gas1 Monoclonal specifically detects Gas1 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Gas1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 309325
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P54826
Gene Alias GAS-1, growth arrest-specific 1, Growth arrest-specific gene-1, growth arrest-specific protein 1
Gene Symbols GAS1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Gas1 Leu40-Ser318 Accession # P54826
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2619
Test Specificity Detects human Gas1 in ELISAs and Western blots. In ELISAs, this antibody does not cross-react with recombinant mouse (rm) Gas1, recombinant human (rh) Gas6, rhArtemin, rhNeurturin, rhRet, or rhShh N-terminal domain. In Western blots, this antibody does not cross-react with rmGas6.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.