Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Gas6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $444.50
Specifications
Antigen | Gas6 |
---|---|
Clone | 100118 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8851SP
![]() |
R&D Systems
MAB8851SP |
25 μg |
Each for $200.50
|
|
|||||
MAB8851100
![]() |
R&D Systems
MAB8851100 |
100 μg |
Each for $444.50
|
|
|||||
Description
Gas6 Monoclonal specifically detects Gas6 in Human samples. It is validated for ELISA.Specifications
Gas6 | |
ELISA | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
AXLLG, AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6 | |
Mouse myeloma cell line NS0-derived recombinant human Gas6, Asp118-Ala678, Accession No. NP_000811 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Gas6 in direct ELISAs. |
100118 | |
ELISA | |
Unconjugated | |
Human | |
NP_000811 | |
2621.0 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title