Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                Human Gas6 Antibody, R&D Systems™
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Mouse Monoclonal Antibody
$200.50 - $414.21
Specifications
| Antigen | Gas6 | 
|---|---|
| Clone | 100130 | 
| Applications | ELISA | 
| Classification | Monoclonal | 
| Conjugate | Unconjugated | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
                
                
                    
                        MAB8853SP
                    
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        R&D Systems
                         MAB8853SP  | 
                
            
            
            
            
            
                
                    25 μg | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $200.50
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
                
                
                    
                        MAB8853100
                    
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        R&D Systems
                         MAB8853100  | 
                
            
            
            
            
            
                
                    100 μg | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $414.21
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
Description
Gas6 Monoclonal specifically detects Gas6 in Human samples. It is validated for ELISA.Specifications
| Gas6 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_000811 | |
| 2621 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | 
| 100130 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AXLLG, AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6 | |
| Mouse myeloma cell line NS0-derived recombinant human Gas6, Asp118-Ala678, Accession # NP_000811 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title