Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GASP-1/WFIKKN2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB20701SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB20701SP 25 μg
MAB20701 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB20701SP Supplier R&D Systems Supplier No. MAB20701SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GASP-1/WFIKKN2 Monoclonal specifically detects GASP-1/WFIKKN2 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen GASP-1/WFIKKN2
Applications Immunohistochemistry
Classification Monoclonal
Clone 288518
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8TEU8
Gene Alias GASP1, GASP-1, Growth and differentiation factor-associated serum protein 1, hGASP-1, WAP, follistatin, immunoglobulin, kunitz and NTR domain-containing-relatedprotein, WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2, WAP, FS, Ig, two KU and NTR module related protein, WAP, kazal, immunoglobulin, kunitz and NTR domain-containing protein 2, WFIKKN2, WFIKKN-related protein, WFIKKNRP, WFIKKNRPmultivalent protease inhibitor protein
Gene Symbols WFIKKN2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human GASP-1/WFIKKN2 Leu35-His576 Accession # Q8TEU8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124857
Test Specificity Detects human GASP-1/WFIKKN2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human GASP-2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.