Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GASP-2/WFIKKN Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | GASP-2/WFIKKN |
---|---|
Clone | 289802 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB21363SP
![]() |
R&D Systems
MAB21363SP |
25 μg |
Each for $200.50
|
|
|||||
MAB21363
![]() |
R&D Systems
MAB21363 |
100 μg |
Each for $468.00
|
|
|||||
Description
GASP-2/WFIKKN Monoclonal specifically detects GASP-2/WFIKKN in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
GASP-2/WFIKKN | |
LYOPH | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
Q96NZ8 | |
117166 | |
Mouse myeloma cell line NS0-derived recombinant human GASP-2/WFIKKN Ala20-Asp548 Accession # Q96NZ8 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human GASP-2/WFIKKN in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human GASP-1 is observed. |
289802 | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C16orf12, Chromosome 16 Open Reading Frame 12, GASP2, GASP-2, Growth And Differentiation Factor-Associated Serum Protein 2, hGASP-2, RJD2, WAP Four-Disulfide Core Domain 20A, WAP, Follistatin/Kazal, Immunoglobulin, Kunitz And Netrin Domain Containing 1, WAP, FS, Ig, KU, And NTR-Containing Protein, WFDC20A, WFIKKN, WFIKKN1 | |
WFIKKN1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title