Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Gastrokine 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6395SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6395SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6395SP Supplier R&D Systems Supplier No. MAB6395SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Gastrokine 1 Monoclonal specifically detects Gastrokine 1 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Gastrokine 1
Applications Immunohistochemistry
Classification Monoclonal
Clone 665701
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9NS71
Gene Alias 18 kDa antrum mucosa protein, AMP18, AMP-18, AMP18BRICHOS domain containing 1, BRICD1, CA11, CA11MGC70354, FOV, foveolin, gastrokine 1, gastrokine-1, GKN1, Protein CA11
Gene Symbols GKN1
Host Species Mouse
Immunogen E. coli-derived recombinant human Gastrokine 1 Asn35-Asn199 Accession # Q9NS71
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56287
Test Specificity Detects human Gastrokine 1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Gastrokine 1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.