Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GATA-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1779SP
This item is not returnable.
View return policy
Description
GATA-1 Polyclonal specifically detects GATA-1 in Human samples. It is validated for Western Blot.Specifications
GATA-1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Eryf1, ERYF1GATA-binding protein 1 (globin transcription factor 1), GATA binding protein 1 (globin transcription factor 1), GATA1, GATA-1erythroid transcription factor, GATA-binding factor 1, GF-1, GF1globin transcription factor 1, NF-E1 DNA-binding protein, NFE1erythroid transcription factor 1, transcription factor GATA1, XLTT | |
Goat | |
Affinity Purified | |
RUO | |
2623 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
P15976 | |
GATA1 | |
E. coli-derived recombinant human GATA-1 Met1-Ser413 Accession # P15976 | |
25 μg | |
Primary | |
Detects human GATA-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows less than 1% cross-reactivity with rhGATA-2, rhGATA-5 and rhGATA-6. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction