Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GATA-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1779SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1779SP 25 μg
MAB1779 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1779SP Supplier R&D Systems Supplier No. MAB1779SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

GATA-1 Monoclonal specifically detects GATA-1 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen GATA-1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 234732
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P15976
Gene Alias Eryf1, ERYF1GATA-binding protein 1 (globin transcription factor 1), GATA binding protein 1 (globin transcription factor 1), GATA1, GATA-1erythroid transcription factor, GATA-binding factor 1, GF-1, GF1globin transcription factor 1, NF-E1 DNA-binding protein, NFE1erythroid transcription factor 1, transcription factor GATA1, XLTT
Gene Symbols GATA1
Host Species Rat
Immunogen E. coli-derived recombinant human GATA-1 Met1-Ser413 Accession # P15976
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2623
Test Specificity Detects human GATA-1 in direct ELISAs. In Western blots, this antibody does not cross-react with rhGATA-2 or rhGATA-6.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.