Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GATA-1 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC1779P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1779P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC1779P Supplier R&D Systems Supplier No. IC1779P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

GATA-1 Monoclonal specifically detects GATA-1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen GATA-1
Applications Flow Cytometry
Classification Monoclonal
Clone 234732
Conjugate R-PE
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P15976
Gene Alias Eryf1, ERYF1GATA-binding protein 1 (globin transcription factor 1), GATA binding protein 1 (globin transcription factor 1), GATA1, GATA-1erythroid transcription factor, GATA-binding factor 1, GF-1, GF1globin transcription factor 1, NF-E1 DNA-binding protein, NFE1erythroid transcription factor 1, transcription factor GATA1, XLTT
Gene Symbols GATA1
Host Species Rat
Immunogen E. coli-derived recombinant human GATA-1 Met1-Ser413 Accession # P15976
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2623
Test Specificity Detects human GATA-1 in direct ELISAs. In Western blots, this antibody does not cross-react with recombinant human (rh) GATA-2 or rhGATA-6.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.