Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GATA-4 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF2606
This item is not returnable.
View return policy
Description
GATA-4 Polyclonal specifically detects GATA-4 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
GATA-4 | |
Polyclonal | |
Biotin | |
P43694 | |
GATA4 | |
E. coli-derived recombinant human GATA-4 Met27-Phe211 Accession # P43694 | |
50 μg | |
Primary | |
Detects human GATA-4 in Western blots. In Western blots, less than 2% cross-reactivity with recombinant human GATA-1, -2, -3, -5, and -6 is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
GATA binding protein 4, GATA4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4 | |
Goat | |
Affinity Purified | |
RUO | |
2626 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction