Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GATA-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB1700
This item is not returnable.
View return policy
Description
GATA-6 Monoclonal specifically detects GATA-6 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| GATA-6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| CAA64997 | |
| GATA6 | |
| E. coli-derived recombinant human GATA-6 Met1-Ala449 Accession # CAA64997 | |
| 100 μg | |
| Primary | |
| Detects human GATA-6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) GATA-1 or rhGATA-2. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 222228 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GATA binding protein 6, GATA6, GATA-binding factor 6, GATA-binding protein 6, transcription factor GATA-6 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2627 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction