Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GCHFR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7918SP
This item is not returnable.
View return policy
Description
GCHFR Monoclonal specifically detects GCHFR in Human samples. It is validated for Western Blot, Simple Western.Specifications
GCHFR | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
P30047 | |
GCHFR | |
E. coli-derived recombinant human GCHFR Met1-Glu84 Accession # P30047 | |
25 μg | |
Primary | |
Detects human GCHFR in ELISAs. | |
Human | |
Purified |
Western Blot | |
846011 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
GFRP, GFRPP35, GTP cyclohydrolase I feedback regulator, GTP cyclohydrolase I feedback regulatory proteinGTP cyclohydrolase 1 feedback regulatory protein, HsT16933, MGC138467, MGC138469, p35 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2644 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction