Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GCN2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6878
This item is not returnable.
View return policy
Description
GCN2 Monoclonal specifically detects GCN2 in Human samples. It is validated for Western Blot.Specifications
GCN2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9P2K8 | |
EIF2AK4 | |
E. coli-derived recombinant human GCN2 Arg22-Lys139 Accession # Q9P2K8 | |
100 μg | |
Primary | |
Detects human GCN2 in direct ELISAs. | |
Human | |
Purified |
Western Blot | |
708210 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EIF2AK4, eukaryotic translation initiation factor 2 alpha kinase 4, eukaryotic translation initiation factor 2-alpha kinase 4, GCN2GCN2 eIF2alpha kinase, GCN2-like protein, KIAA1338EC 2.7.11.1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
440275 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction