Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GCNF/NR6A1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3180
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3180 100 μg
AF3180SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3180 Supplier R&D Systems Supplier No. AF3180
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

GCNF/NR6A1 Polyclonal specifically detects GCNF/NR6A1 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen GCNF/NR6A1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q15406
Gene Alias GCNF1, GCNFNR61, Germ cell nuclear factor, hGCNF, NR6A1, nuclear receptor subfamily 6 group A member 1, nuclear receptor subfamily 6, group A, member 1, retinoic acid receptor-related testis-associated receptor, Retinoid receptor-related testis-specific receptor, RTR, RTRhRTR
Gene Symbols NR6A1
Host Species Goat
Immunogen E. coli-derived recombinant human GCNF/NR6A1 Met215-Glu480 Accession # Q15406
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2649
Test Specificity Detects human and mouse GCNF/NR6A1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.