Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GDF-11/BMP-11 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB19581SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB19581SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB19581SP Supplier R&D Systems Supplier No. MAB19581SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 9 publications

GDF-11/BMP-11 Monoclonal specifically detects GDF-11/BMP-11 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen GDF-11/BMP-11
Applications Immunohistochemistry
Classification Monoclonal
Clone 743833
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O95390
Gene Alias BMP11, BMP-11, BMP-11BMP11Bone morphogenetic protein 11, GDF11, GDF-11, growth differentiation factor 11, growth/differentiation factor 11
Gene Symbols GDF11
Host Species Mouse
Immunogen E. coli-derived recombinant human GDF-11/BMP-11 Asn299-Ser407 Accession # O95390
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10220
Test Specificity Detects human GDF-11/BMP-11 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human BMP-6 or recombinant mouse GDF-8 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.