Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GDF-11/BMP-11 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.50
Specifications
Antigen | GDF-11/BMP-11 |
---|---|
Clone | 937320 |
Dilution | Immunocytochemistry 8-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB19583SP
![]() |
R&D Systems
MAB19583SP |
25 μg |
Each for $200.50
|
|
|||||
MAB19583
![]() |
R&D Systems
MAB19583 |
100 μg |
Each for $468.50
|
|
|||||
Description
GDF-11/BMP-11 Monoclonal specifically detects GDF-11/BMP-11 in Human samples. It is validated for Immunocytochemistry.Specifications
GDF-11/BMP-11 | |
Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BMP11, BMP-11, BMP-11BMP11Bone morphogenetic protein 11, GDF11, GDF-11, growth differentiation factor 11, growth/differentiation factor 11 | |
GDF11 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
937320 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
O95390 | |
10220 | |
E. coli-derived recombinant human GDF-11/BMP-11 Asn299-Ser407 Accession # O95390 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human GDF-11/BMP-11 in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title