Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GDF-15 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | GDF-15 |
---|---|
Clone | 147640 |
Dilution | Immunocytochemistry 8-25 μg/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9571SP
![]() |
R&D Systems
MAB9571SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9571100
![]() |
R&D Systems
MAB9571100 |
100 μg |
Each for $467.50
|
|
|||||
Description
GDF-15 Monoclonal specifically detects GDF-15 in Human samples. It is validated for Immunocytochemistry.Specifications
GDF-15 | |
Immunocytochemistry 8-25 μg/mL | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
GDF15, growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDF, PDFGDF-15, PLAB, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGF-beta, PTGFBPTGF-beta | |
Chinese hamster ovary cell line CHO-derived recombinant human GDF-15 Ala197-Ile308 Accession # Q99988 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human GDF-15 in direct ELISAs. |
147640 | |
Immunocytochemistry | |
Unconjugated | |
Human | |
Q99988 | |
9518 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title