Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GDF-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | GDF-3 |
---|---|
Clone | 878751 |
Dilution | Neutralization 7.5-15 ug/mL |
Applications | Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB57541SP
![]() |
R&D Systems
MAB57541SP |
25 μg |
Each for $200.50
|
|
|||||
MAB57541
![]() |
R&D Systems
MAB57541 |
100 μg |
Each for $491.00
|
|
|||||
Description
GDF-3 Monoclonal specifically detects GDF-3 in Human samples. It is validated for Neutralization.Specifications
GDF-3 | |
Neutralization 7.5-15 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
GDF3, GDF-3, growth differentiation factor 3, growth/differentiation factor 3, KFS3, MCOP7, MCOPCB6, Vgr-2 | |
GDF3 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
878751 | |
Neutralization | |
Unconjugated | |
Mouse | |
Human | |
Q9NR23 | |
9573 | |
Chinese hamster ovary cell line CHO-derived human recombinant GDF-3/BMP-2 heterodimer Ala251-Gly364 Accession # Q9NR23 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human GDF-3 in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) BMP-2 and rhGDF-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title