Learn More
Gibco™ Human GDF-3 Recombinant Protein, PeproTech®

Description
Recombinant Human GDF-3 is a 26.0 kDa non-disulfide-linked homodimer containing two 114 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Specifications
Specifications
Accession Number | Q9NR23 |
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 9573 |
Molecular Weight (g/mol) | 26 kDa |
Name | Human GDF-3 |
Quantity | 100 μg |
Source | E. coli |
Regulatory Status | RUO |
Endotoxin Concentration | <1 EU/ μg |
Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.