Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GDF-5/BMP-14 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8340SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8340SP 25 μg
MAB8340100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8340SP Supplier R&D Systems Supplier No. MAB8340SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GDF-5/BMP-14 Monoclonal antibody specifically detects GDF-5/BMP-14 in Human samples. It is validated for Immunocytochemistry

Specifications

Antigen GDF-5/BMP-14
Applications Immunocytochemistry
Classification Monoclonal
Clone 1065032
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene Accession No. P43026
Gene Alias BMP14, BMP-14, Cartilage-derived morphogenetic protein 1, cartilage-derived morphogenetic protein-1, CDMP-1, CDMP1LAP4, GDF5, GDF-5, growth differentiation factor 5, growth/differentiation factor 5, OS5, Radotermin, SYNS2
Host Species Mouse
Immunogen E. coli-derived recombinant human GDF-5, Ala382-Arg501, Accession # P43026
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8200
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.