Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GDNF Antibody, R&D Systems™

Mouse Monoclonal Antibody
$201.00 - $526.50
Specifications
Antigen | GDNF |
---|---|
Clone | 27106R |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 μg;/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB212RSP
![]() |
R&D Systems
MAB212RSP |
25 μg |
Each for $201.00
|
|
|||||
MAB212R100
![]() |
R&D Systems
MAB212R100 |
100 μg |
Each for $526.50
|
|
|||||
Description
GDNF Monoclonal specifically detects GDNF in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
GDNF | |
ELISA Capture (Matched Antibody Pair) 2-8 μg;/mL | |
Monoclonal | |
Supernatant | |
Human | |
P39905 | |
2668.0 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
27106R | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3 | |
E. coli-derived recombinant human GDNF, Arg109-Ile211, Accession No. P39905 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human GDNF in direct ELISA. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title