Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GDNF Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems BAF212
This item is not returnable.
View return policy
Description
GDNF Polyclonal specifically detects GDNF in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
GDNF | |
Polyclonal | |
Biotin | |
Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3 | |
Goat | |
Affinity Purified | |
RUO | |
2668 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
GDNF | |
recombinant human GDNF | |
50 μg | |
Primary | |
Detects human GDNF in ELISAs and Western blots. In sandwich immunoassays, less than 30% cross-reactivity with recombinant rat GDNF is observed and less than 0.1% cross-reativity with recombinant human (rh) BDNF, rhCNTF, rhNeurturin, rh beta-NGF, rhNT-3, and rhNT-4 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction