Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GDPD5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10524100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10524100 100 μg
MAB10524SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10524100 Supplier R&D Systems Supplier No. MAB10524100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GDPD5 Monoclonal specifically detects GDPD5 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen GDPD5
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 506020
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. Q8WTR4
Gene Alias EC 3.1, GDE2, GDE2PP1665Glycerophosphodiester phosphodiesterase 2, glycerophosphodiester phosphodiesterase domain containing 5, glycerophosphodiester phosphodiesterase domain-containing protein 5, PP1665
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived transfected with GDPD5, Accession # Q8WTR4
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81544
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.