Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Gelsolin/GSN ELISA Kit (Colorimetric)
SDP

Catalog No. NB398198 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398198 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398198 Supplier Novus Biologicals™ Supplier No. NBP282209
Add to Cart
Add to Cart

Kit (Colorimetric)

Specifications

Assay Range 6.25 to 400 ng/mL
Assay Sensitivity 3.75 ng/mL
Immunoassay Kit Format Colorimetric Detection Kit
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Cell Supernatant
Specificity This kit recognizes Human Gelsolin/GSN in samples. No Significant cross-reactivity or interference between Human Gelsolin/GSN and analogues was observed.
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 2934
Gene Symbol GSN
Quantity 1 Kit
Research Discipline Apoptosis, Cancer, Cell Biology, Cellular Markers, Tumor Suppressors
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym Actin-depolymerizing factor, ADF, AGEL, Brevin, DKFZp313L0718, gelsolin, gelsolin (amyloidosis, Finnish type)
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.