Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GFAP Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $526.00
Specifications
Antigen | GFAP |
---|---|
Clone | 987268 |
Dilution | Western Blot 0.25 ug/mL, Immunohistochemistry 0.2-25 ug/mL, Immunocytochemistry 1-25 ug/mL, Knockout Validated 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoassay |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB25941SP
![]() |
R&D Systems
MAB25941SP |
25 μg |
Each for $200.50
|
|
|||||
MAB25941100
![]() |
R&D Systems
MAB25941100 |
100 μg |
Each for $526.00
|
|
|||||
Description
GFAP Monoclonal specifically detects GFAP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Knockout Validated.Specifications
GFAP | |
Western Blot 0.25 ug/mL, Immunohistochemistry 0.2-25 ug/mL, Immunocytochemistry 1-25 ug/mL, Knockout Validated 8-25 ug/mL | |
Monoclonal | |
Supernatant | |
Human | |
P14136 | |
2670 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
50 |
987268 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoassay | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
FLJ45472, GFAP astrocytes, glial fibrillary acidic protein | |
Synthetic peptide containing human GFAP Accession # P14136 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human GFAP in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title