Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GFR alpha-1/GDNF R alpha-1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF714
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF714 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF714 Supplier R&D Systems Supplier No. BAF714
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

GFR alpha-1/GDNF R alpha-1 Polyclonal specifically detects GFR alpha-1/GDNF R alpha-1 in Human samples. It is validated for Western Blot.

Specifications

Antigen GFR alpha-1/GDNF R alpha-1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. NP_665736
Gene Alias DKFZp313E1012, DKFZp686J0156, GDNF family receptor alpha 1, GDNF family receptor alpha-1, GDNF R alpha-1, GDNF receptor alpha-1, GDNFRAGFR-ALPHA-1, GDNFR-alpha-1, GDNFRMGC23045, GFR alpha1, GFRa-1, GFRA1, GFR-alpha-1, Glial cell line-derived neurotrophic factor receptor alpha, GPI-linked anchor protein, PI-linked cell-surface accessory protein, RET ligand 1, RET1L, RETL1FLJ10561, TGF-beta related neurotrophic factor receptor 1, TGF-beta-related neurotrophic factor receptor 1, TRNR1FLJ31546
Gene Symbols GFRA1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human GFR alpha-1/GDNF R alpha-1 Met1-Lys429 Accession # NP_665736
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2674
Test Specificity Detects human GFR alpha-1/GDNF R alpha-1 in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant rat GFR alpha-1 is observed and less than 2% cross-reactivity with recombinant human (rh) GFR alpha-2, rhGFR alpha-3, and rhGFR alpha-4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.