Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GFR alpha-2/GDNF R alpha-2 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Catalog No. FAB6131N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6131N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6131N100 Supplier R&D Systems Supplier No. FAB6131N100UG
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GFR alpha-2/GDNF R alpha-2 Monoclonal specifically detects GFR alpha-2/GDNF R alpha-2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen GFR alpha-2/GDNF R alpha-2
Applications Flow Cytometry
Classification Monoclonal
Clone 71515
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. O00451
Gene Alias GDNF family receptor alpha 2, GDNF family receptor alpha-2, GDNF R alpha-2, GDNF receptor alpha-2, GDNF receptor beta, GDNFR-alpha-2, GDNFR-beta, GDNFRBRET ligand 2, GFR alpha2, GFRa-2, GFRA2, GFR-alpha 2, GFR-alpha-2, glial cell line derived neurotrophic factor receptor, beta, Neurturin receptor alpha, NRTNR-alpha, NTNRA, NTNR-alpha, PI-linked cell-surface accessory protein, RETL2TGF-beta-related neurotrophic factor receptor 2, TGF-beta related neurotrophic factor receptor 2, TRN receptor, GPI-anchored, TRNR2NRTNR-ALPHA
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human GFR alpha-2/GDNF R alpha-2, Ser22-Ser441, Accession # O00451
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2675
Test Specificity Detects human GFR alpha-2/GDNF R alpha-2 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.