Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human GFR alpha-2/GDNF R alpha-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB613
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB613 500 μg
MAB613SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB613 Supplier R&D Systems Supplier No. MAB613
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GFR alpha-2/GDNF R alpha-2 Monoclonal specifically detects GFR alpha-2/GDNF R alpha-2 in Human samples. It is validated for Western Blot.

Specifications

Antigen GFR alpha-2/GDNF R alpha-2
Applications Western Blot
Classification Monoclonal
Clone 129030
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O00451
Gene Alias GDNF family receptor alpha 2, GDNF family receptor alpha-2, GDNF R alpha-2, GDNF receptor alpha-2, GDNF receptor beta, GDNFR-alpha-2, GDNFR-beta, GDNFRBRET ligand 2, GFR alpha2, GFRa-2, GFRA2, GFR-alpha 2, GFR-alpha-2, glial cell line derived neurotrophic factor receptor, beta, Neurturin receptor alpha, NRTNR-alpha, NTNRA, NTNR-alpha, PI-linked cell-surface accessory protein, RETL2TGF-beta-related neurotrophic factor receptor 2, TGF-beta related neurotrophic factor receptor 2, TRN receptor, GPI-anchored, TRNR2NRTNR-ALPHA
Gene Symbols GFRA2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human GFR alpha-2/GDNF R alpha-2 Ser22-Ser441 Accession # O00451
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2675
Test Specificity Detects human GFR alpha-2/GDNF R alpha-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) GDNF R alpha , rhGFR alpha-3, rhGFRa-4, and recombinant mouse GFR alpha-2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.