Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GFR alpha-2/GDNF R alpha-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $468.50
Specifications
Antigen | GFR alpha-2/GDNF R alpha-2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction 2-6 ug/mL |
Applications | Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction |
Classification | Polyclonal |
Description
GFR alpha-2/GDNF R alpha-2 Polyclonal specifically detects GFR alpha-2/GDNF R alpha-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.Specifications
GFR alpha-2/GDNF R alpha-2 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction 2-6 ug/mL | |
Polyclonal | |
Goat | |
Human | |
GDNF family receptor alpha 2, GDNF family receptor alpha-2, GDNF R alpha-2, GDNF receptor alpha-2, GDNF receptor beta, GDNFR-alpha-2, GDNFR-beta, GDNFRBRET ligand 2, GFR alpha2, GFRa-2, GFRA2, GFR-alpha 2, GFR-alpha-2, glial cell line derived neurotrophic factor receptor, beta, Neurturin receptor alpha, NRTNR-alpha, NTNRA, NTNR-alpha, PI-linked cell-surface accessory protein, RETL2TGF-beta-related neurotrophic factor receptor 2, TGF-beta related neurotrophic factor receptor 2, TRN receptor, GPI-anchored, TRNR2NRTNR-ALPHA | |
GFRA2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
Unconjugated | |
RUO | |
O00451 | |
2675 | |
Mouse myeloma cell line NS0-derived recombinant human GFR alpha-2/GDNF R alpha-2 Ser22-Ser441 Accession # O00451 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human GFR alpha-2/GDNF R alpha-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 75% cross-reactivity with recombinant mouse GFR alpha-2 is observed and less than 5% cross-reactivity with recombinant human GFR alpha-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title