Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GFR alpha-like Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB9697G100UG
This item is not returnable.
View return policy
Description
GFR alpha-like Monoclonal specifically detects GFR alpha-like in Human samples. It is validated for Flow Cytometry.Specifications
GFR alpha-like | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Human | |
Supernatant |
Flow Cytometry | |
2345C | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q6UXV0 | |
Human embryonic kidney cell line HEK293-derived recombinant human GFR alpha-like, Ser19-Glu351, Accession # Q6UXV0 | |
100 μg | |
Detects human GFR alpha-like in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction